Learn More
Biomatik Corporation Rat Endothelial protein C receptor (EPCR) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11244
Encompass_Preferred
Rat Endothelial protein C receptor (EPCR) Mini sample ELISA Kit for detecting EPCR in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4V8I1. Gene ID: 362248. Target Synonyms: CD201; PROCR; CCCA ;CCD41; BA42O4.2; EPCR; Endothelial Protein C Receptor; Cell Cycle, Centrosome-Associated Protein; Activated Protein C Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5738