Learn More
Sino Biological Anti-Enolase 1/ENO1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80578T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat ENO1 / Enolase 1 / alpha-enolase (Catalog#80578-R07E; NP_036686.2; Met1-Lys434). ENO1 / Enolase 1 / alpha-enolase specific IgG was purified by Rat ENO1 / Enolase 1 / alpha-enolase affinity chromatography.
Catalog No. 50-240-9000