Learn More
American Research Products Inc Rat EotaXin 1 EotaXin 1 CCL11
Catalog No. 5000328841
Encompass
Rat EotaXin 1 (EotaXin 1/CCL11/ECF) ELISA Kit from CUSABIO Detection Range 15 6 pg/ml -1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28841 Supplier American Research Products Inc Supplier No. CSBE07319R
Rat EotaXin 1 (EotaXin 1/CCL11/ECF) ELISA Kit from CUSABIO Detection Range 15 6 pg/ml -1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich