Learn More
Sino Biological Anti-EpCAM/TROP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80306T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat EpCAM / TROP-1 / TACSTD1 (Catalog#80306-R08H; O55159; Met1-Thr266). EpCAM / TROP-1 / TACSTD1 specific IgG was purified by Rat EpCAM / TROP-1 / TACSTD1 affinity chromatography.
Catalog No. 50-241-2332