Learn More
Sino Biological Anti-EphA7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80109T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat EphA7 (rR EphA7; Catalog# 80109-R08H; Met1-Ser539). EphA7 specific IgG was purified by Rat Cystatin C affinity chromatography.
Catalog No. 50-255-2826