Learn More
Sino Biological Anti-EphB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 81593T36100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat EphB3 / HEK2 / Eph Receptor B3 (rR EphB3 / HEK2 / Eph Receptor B3; Catalog#81593-R08H; NP_001099338.1; Met1-Thr537). EphB3 / HEK2 / Eph Receptor B3 specific IgG was purified by Rat EphB3 / HEK2 / Eph Receptor B3 affinity chromatography.
Catalog No. 50-240-7680