Learn More
Sino Biological Recombinant Rat EphB3 Protein (ECD, His Tag)
Supplier:  Sino Biological 81593R08H200
Encompass_Preferred
A DNA sequence encoding the rat Ephb3 (NP_001099338.1) (Met1-Thr537) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-4142