Learn More
American Research Products Inc Rat Epidermal growth factor E
Catalog No. 5000328800
Encompass
Rat Epidermal growth factor EGF ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28800 Supplier American Research Products Inc Supplier No. CSBE08029R
Rat Epidermal growth factor EGF ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich