Learn More
Biomatik Corporation Rat Epidermal Growth Factor (EGF) CLIA Kit
Supplier:  Biomatik Corporation EKU08895
Encompass_Preferred
Rat Epidermal Growth Factor (EGF) CLIA Kit for detecting EGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.94pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P07522. Gene ID: 25313. Target Synonyms: URG; Beta-Urogastrone; Pro-epidermal growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8600