Learn More
American Research Products Inc Rat Epinephrine Adrenaline EP
Catalog No. 5000329737
Encompass
Rat Epinephrine/Adrenaline EPI ELISA Kit from CUSABIO Detection Range 1 pg/mL-200 pg/mL Sensitivity 0 5 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29737 Supplier American Research Products Inc Supplier No. CSBE08678R
Rat Epinephrine/Adrenaline EPI ELISA Kit from CUSABIO Detection Range 1 pg/mL-200 pg/mL Sensitivity 0 5 pg/mL Sample serum plasma tissue homogenates Method Competitive