Learn More
Biomatik Corporation Rat Epiregulin (EREG) CLIA Kit
Supplier:  Biomatik Corporation EKU09706
Encompass_Preferred
Rat Epiregulin (EREG) CLIA Kit for detecting EREG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.48pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9Z0L5. Gene ID: 59325. Target Synonyms: EPR; ER; Proepiregulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7271