Learn More
American Research Products Inc Rat Epithelial neutrophil acti
Catalog No. 5000329017
Encompass
Rat Epithelial neutrophil actiVating peptide 78 ENA-78 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29017 Supplier American Research Products Inc Supplier No. CSBE08179R
Rat Epithelial neutrophil actiVating peptide 78 ENA-78 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich