Learn More
Biomatik Corporation Rat Epithelial Neutrophil Activating Peptide 78 (ENA78) CLIA Kit
Supplier:  Biomatik Corporation EKU09656
Encompass_Preferred
Rat Epithelial Neutrophil Activating Peptide 78 (ENA78) CLIA Kit for detecting ENA78 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.49pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P97885. Gene ID: 60665. Target Synonyms: CXCL5; SCYB5; Chemokine C-X-C-Motif Ligand 5; Small Inducible Cytokine Subfamily B (Cys-X-Cys) ,Member 5; Neutrophil-activating peptide ENA-78. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6339