Learn More
Raybiotech Inc Rat Erythropoietin ELISA Kit
Supplier:  Raybiotech Inc ELREPO1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.07 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects rat EPO and mouse EPO. Other species not determined. Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: rat 4-1BB (TNFRSF9/CD137), Activin A, Adiponectin (ACRP30), CD80 (B7-1), CD48 (SLAMF2), CTACK (CCL27), Decorin, Eotaxin-1, EphA5, gp130, HGF, IL-1 R6 (IL-1 Rrp2), IL-1 ra, IL-2 Ralpha, IL-3, IL-7, IL-17F, IL-22, JAM-A (CD321/F11R), MIP-1 alpha, Neuropilin-1, Neuropilin-2, Nope, Notch-1, Notch-2, P-Cadherin, Prolactin, RANTES, SCF, TIM-1 (KIM-1), TIMP-2, TREM-1, TWEAK R.
Catalog No. 50-195-0237