Learn More
BIOHIPPO Rat EPX Microsample
SDP
Catalog No. 5000315812
Encompass
Rat EPX Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EPX in serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 3 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat EPX Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat EPX Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat EPX biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15812 Supplier BIOHIPPO Supplier No. BHE15210557
Rat EPX Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EPX in serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 3 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat EPX Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat EPX Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat EPX biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color