Learn More
Sino Biological Anti-HER3/ERBB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80111RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat HER3 / ErbB3 (Catalog#80111-R08H; Q3MHC0; Met1-His641). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2174