Learn More
BIOHIPPO Rat EREG Microsample
SDP
Catalog No. 5000315171
Encompass
Rat EREG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EREG in Serum plasma tissue homogenates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat EREG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat EREG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat EREG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15171 Supplier BIOHIPPO Supplier No. BHE15211019
Rat EREG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EREG in Serum plasma tissue homogenates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat EREG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat EREG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat EREG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color