Learn More
Crystal Chem Inc Rat Estradiol ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80548
Encompass
The Rat Estradiol ELISA is used to quantify levels of rat estradiol in serum using only a 75 µL sample volume. Highlights: Kit uses sample size of only 75 µL. Dynamic range of 5 - 1280 pg/mL. No extraction needed, ready to use reagents. Highly precise with low CV's. High sensitivity of 2.5 pg/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7724