Learn More
Biomatik Corporation Rat Eukaryotic Translation Elongation Factor 1 Alpha 1 (EEF1a1) ELISA Kit
Supplier:  Biomatik Corporation EKU12420
Encompass_Preferred
Rat Eukaryotic Translation Elongation Factor 1 Alpha 1 (EEF1a1) ELISA Kit for detecting EEF1a1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62630. Gene ID: 171361. Target Synonyms: CCS-3; CCS3; EEF-1; EEF1A; EF-Tu; EF1A; FLJ25721; GRAF-1EF; LENG7; PTI1; eEF1A-1; Leukocyte Receptor Cluster (LRC) Member 7; Elongation factor Tu. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5615