Learn More
American Research Products Inc Rat eXtracellular superoXide d
Catalog No. 5000330203
Encompass
Rat eXtracellular superoXide dismutase [Cu-Zn] (Cu/Zn-SOD/SOD3) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-30203 Supplier American Research Products Inc Supplier No. CSBE14981R
Rat eXtracellular superoXide dismutase [Cu-Zn] (Cu/Zn-SOD/SOD3) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich