Learn More
BIOHIPPO Rat F12 Microsample
SDP
Catalog No. 5000314590
Encompass
Rat F12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F12 in plasma It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 26 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat F12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat F12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat F12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14590 Supplier BIOHIPPO Supplier No. BHE15212451
Rat F12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F12 in plasma It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 26 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat F12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat F12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat F12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color