Learn More
Sino Biological Rat F2 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 80722R08H50
Encompass_Preferred
A DNA sequence encoding the rat F2 (NP_075213.2) (Met1-Arg617) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1515