Learn More
Sino Biological Anti-Tissue Factor Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80407MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat Coagulation Factor III / Tissue Factor / CD142 (rR Coagulation Factor III / Tissue Factor / CD142; Catalog#80407-R08H; Q66HI4; Met1-Glu252). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0271