Learn More
Sino Biological Anti-Tissue Factor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80407T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Coagulation Factor III / Tissue Factor / CD142 (Catalog#80407-R08H; Q66HI4; Met1-Glu252). Coagulation Factor III / Tissue Factor / CD142 specific IgG was purified by Rat Coagulation Factor III / Tissue Factor / CD142 affinity chromatography.
Catalog No. 50-241-1864