Learn More
BIOHIPPO Rat F7 Microsample
SDP
Catalog No. 5000314128
Encompass
Rat F7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F7 in Plasma It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 12 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat F7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat F7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat F7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14128 Supplier BIOHIPPO Supplier No. BHE15212506
Rat F7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of F7 in Plasma It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 12 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat F7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat F7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat F7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color