Learn More
American Research Products Inc Rat Factor related Apoptosis l
Catalog No. 5000326462
Encompass
Rat Factor-related Apoptosis ligand FASL ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26462 Supplier American Research Products Inc Supplier No. CSBE07325R
Rat Factor-related Apoptosis ligand FASL ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich