Learn More
BIOHIPPO Rat FAPa Microsample
SDP
Catalog No. 5000315658
Encompass
Rat FAPa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FAPa in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 119 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat FAPa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat FAPa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat FAPa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15658 Supplier BIOHIPPO Supplier No. BHE15210979
Rat FAPa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FAPa in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 119 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat FAPa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat FAPa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat FAPa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color