Learn More
Biomatik Corporation Rat Farnesoid X Receptor (FXR) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11103
Encompass_Preferred
Rat Farnesoid X Receptor (FXR) Mini sample ELISA Kit for detecting FXR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62735. Gene ID: 60351. Target Synonyms: NR1H4; BAR; FXR; HRR1; RIP14; Nuclear Receptor Subfamily 1,Group H,Member 4; Bile acid receptor; Farnesol receptor HRR-1; Retinoid X receptor-interacting protein 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5532