Learn More
Cayman Chemical Rat FarnsoId X RECPTR Repo
SDP
Catalog No. 502940396
Encompass_Preferred
Rat FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00601-32, R00601]
Catalog No. 50-294-0396 Supplier Cayman Chemical Supplier No. 283403X32 WELL
Rat FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00601-32, R00601]