Learn More
Sino Biological Recombinant Anti-Fas/CD95/TNFRSF6 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 80043R028P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD95 / APO-1 / TNFRSF6 (rR CD95 / APO-1 / TNFRSF6; Catalog#80043-R08H; NP_631933.2; Met1-Lys170) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-7777