Learn More
American Research Products Inc Rat Fatty acid amide hydrolase
Catalog No. 5000326879
Encompass
Rat Fatty-acid amide hydrolase 1 (FAAH) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26879 Supplier American Research Products Inc Supplier No. CSBEL007938RA
Rat Fatty-acid amide hydrolase 1 (FAAH) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich