Learn More
Biomatik Corporation Rat Fatty Acid Binding Protein 5 (FABP5) ELISA Kit
Supplier:  Biomatik Corporation EKU10816
Encompass_Preferred
Rat Fatty Acid Binding Protein 5 (FABP5) ELISA Kit for detecting FABP5 in serum, plasma, tissue homogenates, cell lysates, breast milk, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.069ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55053. Gene ID: 140868. Target Synonyms: E-FABP; EFABP; PA-FABP; PAFABP; Fatty Acid Binding Protein 5, Epidermal; Psoriasis-Associated; Epidermal-type fatty acid-binding protein; Psoriasis-associated fatty acid-binding protein homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5265