Learn More
Biomatik Corporation Rat Fatty Acid Binding Protein 6, Ileal (FABP6) ELISA Kit
Supplier:  Biomatik Corporation EKU04044
Encompass_Preferred
Rat Fatty Acid Binding Protein 6, Ileal (FABP6) ELISA Kit for detecting FABP6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80020. Gene ID: 25440. Target Synonyms: GT; iBABP; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ILLBP; I-15P; Gastrotropin; Illeal Lipid-Binding Protein; Ileal Bile Acid Binding Protein; Intestinal 15 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7356