Learn More
American Research Products Inc Rat fatty acid binding protein
Catalog No. 5000326390
Encompass
Rat fatty acid binding protein 4 adipocyte (FABP4) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26390 Supplier American Research Products Inc Supplier No. CSBE13027R
Rat fatty acid binding protein 4 adipocyte (FABP4) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich