Learn More
American Research Products Inc Rat fatty acid synthase FAS
Catalog No. 5000328622
Encompass
Rat fatty acid synthase (FAS) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28622 Supplier American Research Products Inc Supplier No. CSBE16440R
Rat fatty acid synthase (FAS) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich