Learn More
Innovative Research Inc Rat Fbrngn Kit
Catalog No. 502744009
Encompass_Preferred
Rat Fibrinogen ELISA Kit - This rat fibrinogen antigen assay is intended for the quantitative determination of total fibrinogen antigen in rat plasma and serum. For research use only. Strip well format. Reagents for up to 96 tests. Rat fibrinogen will bind to the affinity purified capture antibody coated on the microtiter plate. After appropriate washing steps, biotin labeled polyclonal anti-rat fibrinogen primary antibody binds to the captured protein. Excess antibody is washed away and bound polyclonal antibody is reacted with streptavidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. A standard calibration curve is prepared along with the samples to be measured using dilutions of rat fibrinogen. Color development is proportional to the concentration of fibrinogen in the samples.
Catalog No. 50-274-4009 Supplier Innovative Research Inc Supplier No. IRTFBGKT
Rat Fibrinogen ELISA Kit - This rat fibrinogen antigen assay is intended for the quantitative determination of total fibrinogen antigen in rat plasma and serum. For research use only. Strip well format. Reagents for up to 96 tests. Rat fibrinogen will bind to the affinity purified capture antibody coated on the microtiter plate. After appropriate washing steps, biotin labeled polyclonal anti-rat fibrinogen primary antibody binds to the captured protein. Excess antibody is washed away and bound polyclonal antibody is reacted with streptavidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. A standard calibration curve is prepared along with the samples to be measured using dilutions of rat fibrinogen. Color development is proportional to the concentration of fibrinogen in the samples.