Learn More
ACROBiosystems ACROBiosystems Rat Fc gamma RIIA / CD32a Protein, His Tag
Catalog No. 502106329
Encompass_Preferred
Recombinant Protein;1MG;Rat CD32a, His Tag (CDA-R52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 34 - Leu216 (Accession # M0R4F7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa.
Catalog No. 50-210-6329 Supplier ACROBiosystems Supplier No. CDAR52H51MG
Recombinant Protein;1MG;Rat CD32a, His Tag (CDA-R52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 34 - Leu216 (Accession # M0R4F7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa.