Learn More
Sino Biological Recombinant Rat CD23 Protein (hFc Tag)
Supplier:  Sino Biological 80208R01H1
Encompass_Preferred
A DNA sequence encoding the rat FCER2 (Q6AZ45) (Glu50-Pro331) was expressed with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-255-2811