Learn More
Sino Biological Rat FCGR1 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 80016R005A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD64/Fc gamma RI (rR CD64/Fc gamma RI; Catalog#80016-R08H; NP_001094306.1; Met1-Pro285) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-2624