Learn More
Sino Biological Recombinant Anti-CD64/FCGR1 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 80016R005F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD64 / FCGR1 (rR CD64 / FCGR1; Catalog#80016-R08H; NP_001094306.1; Met1-Pro285) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8883