Learn More
Sino Biological Anti-CD64/FCGR1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80016MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD64 / FCGR1A (rR CD64 / FCGR1A; Catalog #80016-R08H; Met 1-Pro 285; NP_001094306.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2589