Learn More
Sino Biological Anti-CD64/FCGR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80016RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD64 / FCGR1 (Catalog#80016-R08H; NP_001094306.1; Met1-Pro285). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8875