Learn More
Sino Biological Recombinant Rat CD32A/FCGR2A Protein (His Tag)
Supplier:  Sino Biological 80015R08H100
Encompass_Preferred
A DNA sequence encoding the rat FCGR2A (NP_446295.2) (Met1-Leu216) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2876