Learn More
Sino Biological Anti-CD32B/Fcgr2b Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80018RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD32b / Fc gamma RIIB (rR CD32b / Fc gamma RIIB; Catalog#80018-R08H; Q63203-1; Met 1-Pro 212). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8397