Learn More
Sino Biological Recombinant Rat CD32B/Fcgr2b Protein (His Tag)
Supplier:  Sino Biological 80018R08H1
Encompass_Preferred
A DNA sequence encoding the rat FCGR2B (Q63203-1) extracellular domain (Met 1-Pro 212) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2874