Learn More
Sino Biological Recombinant Rat FCGRT & B2M Heterodimer Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological CT030R08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Ser298) of rat FCGRT (P13599) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the rat B2M (P07151) (Met1-Met119) constructed the plasmid 2. The two plasmids were co-expressed and the FCGRT/B2M heterodimer was purified. The purified protein was biotinylated in vitro.
Catalog No. 50-255-2619