Learn More
SAI Infusion Technologies Rodent Oral Gavage (Feeding) Needle - 17G 52mm with Stainless Steel Bite-Guard
SDP
Catalog No. NC1229100
Encompass
17G 52mm Flexible and Disposable Feeding Needles with a Stainless Steel Bite-Guard for maximum durability. SAI's smooth needle tip and flexible PTFE tubing prevent damage to the esophagus during dosing, while the unique Bite-Guard prevents any biting damage to the needle. These needles are sterile and packed 5/pk, 20 packs/box. Product is for laboratory animal research only.
Catalog No. NC1229100 Supplier SAI Infusion Technologies Supplier No. FN1752M
17G 52mm Flexible and Disposable Feeding Needles with a Stainless Steel Bite-Guard for maximum durability. SAI's smooth needle tip and flexible PTFE tubing prevent damage to the esophagus during dosing, while the unique Bite-Guard prevents any biting damage to the needle. These needles are sterile and packed 5/pk, 20 packs/box. Product is for laboratory animal research only.