Learn More
SAI Infusion Technologies Rat Femoral Artery Catheter - 37.8cm PU
SDP
Catalog No. NC3663053
Encompass
Femoral Artery Catheter for Rats. 37.8cm PU catheter with a 7.8cm tip and 4 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle. Product is for laboratory animal research only.
Catalog No. NC3663053 Supplier SAI Infusion Technologies Supplier No. RFACPUX30
Femoral Artery Catheter for Rats. 37.8cm PU catheter with a 7.8cm tip and 4 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle. Product is for laboratory animal research only.