Learn More
SAI Infusion Technologies Rat Femoral Artery Catheter - 125g+, 2.5cm tip
SDP
Supplier:  SAI Infusion Technologies RFAC01
Encompass
This item has a minimum qty of 4 per supplier requirements.
Femoral artery catheter for rat > 125g. 2.5cm 2Fr tip, PU, accepts 22G blunt needle, 2 suture beads to anchor the catheter in place. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply.  Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
Catalog No. NC1397182