Learn More
SAI Infusion Technologies Rat Femoral Vein Catheter - 46cm, for Rats 150g+
SDP
Catalog No. NC0695685
Encompass
Femoral Vein Catheter for Rats 150g or larger. 46cm Tapered PU catheter with a 4cm 2.3Fr tip and 3 suture beads to anchor the catheter in place. Sterile. Access witn a 22G blunt needle. Product is for laboratory animal research only.
Catalog No. NC0695685 Supplier SAI Infusion Technologies Supplier No. RFV04
Femoral Vein Catheter for Rats 150g or larger. 46cm Tapered PU catheter with a 4cm 2.3Fr tip and 3 suture beads to anchor the catheter in place. Sterile. Access witn a 22G blunt needle. Product is for laboratory animal research only.